KEY947335194 Key Organics / BIONET

4-(4-ethylpiperazin-1-yl)benzoic acid

  • MDL number MFCD05864834
  • Cas Number : 784130-66-1
  • Molecular Weight : 234.299

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/404900

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-3029_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart