KEY947335237 Key Organics / BIONET

5-(4-methylpiperazin-1-yl)-1H-1,2,4-triazol-3-amine

  • MDL number MFCD09835389
  • Cas Number : 89292-91-1
  • Molecular Weight : 182.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(4-methyl-1-piperazinyl)-1H-1,2,4-triazol-3-ylamine
Inchi Code 1S/C7H14N6/c1-12-2-4-13(5-3-12)7-9-6(8)10-11-7/h2-5H2,1H3,(H3,8,9,10,11)
Inchi Key OGKMRAGLDFHRGA-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405114

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-3247_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart