KEY947335332 Key Organics / BIONET

methyl 3-(trifluoromethyl)pyridine-2-carboxylate

  • MDL number MFCD14698094
  • Cas Number : 588702-69-6
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-(trifluoromethyl)-2-pyridinecarboxylate
Inchi Code 1S/C8H6F3NO2/c1-14-7(13)6-5(8(9,10)11)3-2-4-12-6/h2-4H,1H3
Inchi Key YRTIWUCFNRNKMA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store under N2
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405465

Safety Information

signalword Danger
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-3607_SDS.pdf
hcodes H226,H300,H311,H331
pictograms GHS02,GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart