KEY947335342 Key Organics / BIONET

3-bromo-7-methylimidazo[1,2-a]pyrimidine

  • MDL number MFCD08234998
  • Cas Number : 375857-62-8
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405502

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-3644_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart