KEY947335375 Key Organics / BIONET

2-(6-fluoro-2,3-dihydro-1H-inden-1-yl)acetic acid

  • MDL number MFCD16303859
  • Cas Number : 1188044-87-2
  • Molecular Weight : 194.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-fluoro-2,3-dihydro-1H-inden-1-yl)acetic acid
Inchi Code 1S/C11H11FO2/c12-9-4-3-7-1-2-8(5-11(13)14)10(7)6-9/h3-4,6,8H,1-2,5H2,(H,13,14)
Inchi Key LHUYQQXFPZPNDL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405601

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-3745_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart