KEY947335428 Key Organics / BIONET

3-ethyl-2-{[(4-fluorophenyl)methyl]sulfanyl}-3,4-dihydroquinazolin-4-one

  • MDL number MFCD03416691
  • Cas Number : 882082-72-6
  • Molecular Weight : 314.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100069

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-2122_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart