KEY947335472 Key Organics / BIONET

5-tert-butyl-2-hydroxy-4'-methoxy-[1,1'-biphenyl]-3-carbaldehyde

  • MDL number MFCD30833907
  • Cas Number : 402576-60-7
  • Molecular Weight : 284.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 119
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/720183

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/NS-00074_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart