KEY947335475 Key Organics / BIONET

methyl (2Z)-2-(chloromethyl)-3-(4-fluorophenyl)prop-2-enoate

  • MDL number MFCD30833910
  • Cas Number : 847653-89-8
  • Molecular Weight : 228.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38 - 40
Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/720187

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/NS-00078_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart