KEY947335481 Key Organics / BIONET

(2Z)-2-iodo-3-[(4-methoxyphenyl)methyl]but-2-ene-1,4-diol

  • MDL number MFCD30833915
  • Cas Number : 2098457-01-1
  • Molecular Weight : 334.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93 - 94
Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/720193

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/NS-00084_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart