KEY947336271 Key Organics / BIONET

5-methyl-1,2-oxazole-3-carboxamide

  • MDL number MFCD00085128
  • Cas Number : 3445-52-1
  • Molecular Weight : 126.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-3-isoxazolecarboxamide
Inchi Code 1S/C5H6N2O2/c1-3-2-4(5(6)8)7-9-3/h2H,1H3,(H2,6,8)
Inchi Key KBOSIRPMGVGOEP-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/459485

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/TS-01656_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart