KEYH042EEBF6 Key Organics / BIONET

ethyl (2E)-3-amino-3-{[4-(trifluoromethoxy)phenyl]amino}prop-2-enoate hydrochloride

  • MDL number MFCD00243911
  • Cas Number : 317821-98-0
  • Molecular Weight : 326.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67174

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2G-903_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart