KEYH042EEBF7 Key Organics / BIONET

N'-cyano-3-fluorobenzene-1-carboximidamide

  • MDL number MFCD27957298
  • Cas Number : 1431559-90-8
  • Molecular Weight : 163.155

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191 - 192
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427456

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-9008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart