KEYH042EEC0A Key Organics / BIONET

4-{[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]methyl}piperidine hydrochloride

  • MDL number MFCD28016126
  • Cas Number : 1239771-50-6
  • Molecular Weight : 297.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432083

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3004_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart