KEYH042EEC0B Key Organics / BIONET

3-(4-methoxyphenyl)-5-(pyrrolidin-2-yl)-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD28016131
  • Cas Number : 853104-22-0
  • Molecular Weight : 281.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432089

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart