KEYH042EEC4F Key Organics / BIONET

Fmoc-Dap(Boc)-OH

  • Cas Number : 198544-42-2
  • Molecular Weight : 426.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R)-3-[(tert-butoxycarbonyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Inchi Code 1S/C23H26N2O6/c1-23(2,3)31-21(28)24-12-19(20(26)27)25-22(29)30-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,12-13H2,1-3H3,(H,24,28)(H,25,29)(H,26,27)/t19-/m1/s1
Inchi Key PKAUMAVONPSDRW-LJQANCHMSA-N

Description

Purity 98%
Storage temp. Store at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/746804

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-10023_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart