KEYH042EF126 Key Organics / BIONET

2,2-Difluoro-2-(fluorosulfonyl)acetic acid

  • Cas Number : 1717-59-5
  • Molecular Weight : 178.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/748040

Safety Information

signalword Danger
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H314,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-11417_SDS.pdf
pictograms GHS05,GHS06,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart