KEYH042EFACE Key Organics / BIONET

Cyproheptadine HCl

  • Cas Number : 969-33-5
  • Molecular Weight : 323.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/750494

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-14697_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart