KEYH042EFDF3 Key Organics / BIONET

4,4'-Bis(hydroxymethyl)-2,2'-bipyridine

  • MDL number MFCD06202808
  • Cas Number : 109073-77-0
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2'-bipyridine-4,4'-diyldimethanol
Inchi Code 1S/C12H12N2O2/c15-7-9-1-3-13-11(5-9)12-6-10(8-16)2-4-14-12/h1-6,15-16H,7-8H2
Inchi Key RRXGRDMHWYLJSY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/751299

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-15762_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart