KEYH042F05BF Key Organics / BIONET

Methyl (R)-1-Boc-piperazine-2-carboxylate

  • MDL number MFCD04115327
  • Cas Number : 252990-05-9
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 4
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/753294

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-18235_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart