KEYH042F0685 Key Organics / BIONET

N-(3-Methylpyridin-2-yl)acetamide

  • Cas Number : 7463-30-1
  • Molecular Weight : 150.181

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/753492

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-18531_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart