KEYH042F0BBC Key Organics / BIONET

2,5-Difluoropyridine-4-boronic acid

  • Cas Number : 1263375-23-0
  • Molecular Weight : 158.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at -18°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/754820

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-20158_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart