KEYH14DE34E8 Key Organics / BIONET

ethyl 2-cyano-3-[3-(trifluoromethoxy)phenyl]prop-2-enoate

  • MDL number MFCD28023499
  • Cas Number : 773089-44-4
  • Molecular Weight : 285.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433301

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10N-919_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart