KEYH14DE34F0 Key Organics / BIONET

ethyl 2-amino-3-[(2-phenylacetyl)sulfanyl]propanoate; trifluoroacetic acid

  • MDL number MFCD06795589
  • Cas Number : 1396965-98-2
  • Molecular Weight : 381.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Hygroscopic
Shipping Temperature Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6270

Safety Information

signalword Warning
pcodes P232, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-098_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart