KEYH14DE34F3 Key Organics / BIONET

N-[3-(dimethylamino)propyl]-4-nitro-2-(trifluoromethyl)aniline hydrochloride

  • MDL number MFCD04035620
  • Cas Number : 1025759-04-9
  • Molecular Weight : 327.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7010

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0659_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart