KEYH14DE34F9 Key Organics / BIONET

N,N-dimethyl-4-({[5-oxo-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-4-ylidene]methyl}amino)benzene-1-sulfonamide

  • MDL number MFCD00138404
  • Cas Number : 320425-21-6
  • Molecular Weight : 362.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63249

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1C-120_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart