KEYH14DE34FA Key Organics / BIONET

N,N-dimethyl-4-{[(5-oxo-3-phenyl-4,5-dihydro-1H-pyrazol-4-ylidene)methyl]amino}benzene-1-sulfonamide

  • MDL number MFCD00138407
  • Cas Number : 320415-75-6
  • Molecular Weight : 370.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63254

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1C-125_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart