KEYH14DE34FB Key Organics / BIONET

4-({[3-(methoxymethyl)-5-oxo-4,5-dihydro-1H-pyrazol-4-ylidene]methyl}amino)-N,N-dimethylbenzene-1-sulfonamide

  • MDL number MFCD00138408
  • Cas Number : 320415-77-8
  • Molecular Weight : 338.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63255

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1C-126_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart