KEYH14DE34FD Key Organics / BIONET

O-[(3-chloro-2-fluorophenyl)methyl]hydroxylamine hydrochloride

  • MDL number MFCD01114580
  • Cas Number : 317821-73-1
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63911

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1G-943_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart