KEYH14DE34FF Key Organics / BIONET

2-(4-methylpiperazin-1-yl)-6-(trifluoromethyl)pyridine-3-carbonitrile hydrochloride

  • MDL number MFCD16628284
  • Cas Number : 166451-06-5
  • Molecular Weight : 306.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64554

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1L-378S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart