KEYH14DE3502 Key Organics / BIONET

4-[2-(benzenesulfonyl)-2-chloroethyl]morpholine hydrochloride

  • MDL number MFCD00232070
  • Cas Number : 317821-96-8
  • Molecular Weight : 326.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66708

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2D-063_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart