KEYH14DE3503 Key Organics / BIONET

N-(2-aminoethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide hydrochloride

  • MDL number MFCD00139577
  • Cas Number : 57005-39-7
  • Molecular Weight : 396.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66795

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2E-115S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart