KEYH14DE3504 Key Organics / BIONET

methyl 2-amino-3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]propanoate hydrochloride

  • MDL number MFCD02661849
  • Cas Number : 1047974-69-5
  • Molecular Weight : 319.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67231

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2H-034_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart