KEYH14DE3508 Key Organics / BIONET

ethyl 2-cyano-3-{4-[(pyridin-2-yl)methoxy]phenyl}prop-2-enoate

  • MDL number MFCD27957297
  • Cas Number : 1565822-18-5
  • Molecular Weight : 308.337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133 - 135
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427454

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-909_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart