KEYH14DE3510 Key Organics / BIONET

4-[(dimethylamino)methylidene]-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00138484
  • Cas Number : 821016-54-0
  • Molecular Weight : 139.158

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243 - 245
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77582

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5B-088_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart