KEYH14DE3516 Key Organics / BIONET

3-(dimethylamino)-1-(4-methoxyphenyl)propan-1-one hydrochloride

  • MDL number MFCD00156682
  • Cas Number : 2125-49-7
  • Molecular Weight : 243.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84435

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6X-0962_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart