KEYH14DE3522 Key Organics / BIONET

3-(dimethylamino)-1-(4-methylphenyl)propan-1-one hydrochloride

  • MDL number MFCD00156711
  • Molecular Weight : 227.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(dimethylamino)-1-(4-methylphenyl)-1-propanone hydrochloride
Inchi Code 1S/C12H17NO.ClH/c1-10-4-6-11(7-5-10)12(14)8-9-13(2)3;/h4-7H,8-9H2,1-3H3;1H
Inchi Key SDNGTEVPTBGTKK-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88210

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7X-0926_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart