KEYH14DE353B Key Organics / BIONET

4-chloro-N'-{[5-oxo-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-4-ylidene]methyl}benzohydrazide

  • MDL number MFCD00138344
  • Cas Number : 338975-58-9
  • Molecular Weight : 332.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91457

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9B-108_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart