KEYH14DE353D Key Organics / BIONET

1-(dimethylamino)-4-phenoxypent-1-en-3-one

  • MDL number MFCD00139117
  • Cas Number : 338976-01-5
  • Molecular Weight : 219.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91510

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9C-064_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart