KEYH14DE354E Key Organics / BIONET

1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}azepane hydrochloride

  • MDL number MFCD23379917
  • Molecular Weight : 351.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/363003

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2613_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart