KEYH14DE3C15 Key Organics / BIONET

2-(3-methylphenoxy)ethylamine

  • MDL number MFCD02856329
  • Cas Number : 6487-99-6
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-methylphenoxy)ethylamine
Inchi Code 1S/C9H13NO/c1-8-3-2-4-9(7-8)11-6-5-10/h2-4,7H,5-6,10H2,1H3
Inchi Key IGGVZLIHUYNMNQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store at 4
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1016099

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-31742_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart