KEYH14DE596D Key Organics / BIONET

4-methoxy-3-(trifluoromethyl)aniline

  • Cas Number : 393-15-7
  • Molecular Weight : 191.153

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1023605

Safety Information

signalword Danger
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335,H412
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-39286_SDS.pdf
pictograms GHS07,GHS09
Availability details for
Quantity
  • Availability
  • Add to Cart