KEYH14DE6CC1 Key Organics / BIONET

1-boc-4-chloroacetylpiperazine

  • Cas Number : 190001-40-2
  • Molecular Weight : 262.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1028554

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-44245_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart