KEYH14DE80A5 Key Organics / BIONET

h2n-peg4-tbu

  • Cas Number : 581065-95-4
  • Molecular Weight : 321.414

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2 at -18°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1033646

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-49352_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart