KEYH14DE8487 Key Organics / BIONET

tert-butyl 2,5-dibromo-1h-pyrrole-1-carboxylate

  • MDL number MFCD11112105
  • Cas Number : 117657-38-2
  • Molecular Weight : 325

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 4
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1034640

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-50351_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart