KEYH14DE8DE5 Key Organics / BIONET

tert-butyl trans-3-hydroxy-4-methylpiperidine-1-carboxylate

  • MDL number MFCD18791214
  • Cas Number : 1932302-38-9
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (3R,4S)-3-hydroxy-4-methyl-1-piperidinecarboxylate
Inchi Code 1S/C11H21NO3/c1-8-5-6-12(7-9(8)13)10(14)15-11(2,3)4/h8-9,13H,5-7H2,1-4H3/t8-,9-/m0/s1
Inchi Key ADHBFIMXBSFQLJ-IUCAKERBSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1037039

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-52762_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart