KEYH14DE9660 Key Organics / BIONET

(r)-2-(trifluoromethyl)oxirane

  • Cas Number : 143142-90-9
  • Molecular Weight : 112.051

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 90
Storage temp. Store at -18°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1039211

Safety Information

signalword Danger
pcodes P260,P270,P280,P402 + P404
hcodes H224,H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-54996_SDS.pdf
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart