KEYH14DEAE71 Key Organics / BIONET

Methyl 5-bromo-2-hydroxyisonicotinate

  • MDL number MFCD09027265
  • Cas Number : 913836-17-6
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-2-oxo-1,2-dihydro-4-pyridinecarboxylate
Inchi Code 1S/C7H6BrNO3/c1-12-7(11)4-2-6(10)9-3-5(4)8/h2-3H,1H3,(H,9,10)
Inchi Key XOMMLGBSSAVOET-UHFFFAOYSA-N

Description

Purity 98%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/854645

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-11194_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart