KEYH14DEC186 Key Organics / BIONET

(R)-2-Benzamido-2-phenylacetic acid

  • MDL number MFCD02684297
  • Cas Number : 10419-67-7
  • Molecular Weight : 255.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R)-(benzoylamino)(phenyl)ethanoic acid
Inchi Code 1S/C15H13NO3/c17-14(12-9-5-2-6-10-12)16-13(15(18)19)11-7-3-1-4-8-11/h1-10,13H,(H,16,17)(H,18,19)/t13-/m1/s1
Inchi Key ACDLFRQZDTZESK-CYBMUJFWSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/859530

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-16147_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart