KEYH14DEC300 Key Organics / BIONET

3-(4-Nitro-1-oxoisoindolin-2-yl)piperidine-2,6-dione

  • Cas Number : 827026-45-9
  • Molecular Weight : 289.247

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/859908

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DS-16532_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart