KEYH14DEC89F Key Organics / BIONET

5-Cyano-2-fluorobenzene-1-sulfonyl chloride

  • MDL number MFCD18394026
  • Cas Number : 1101120-80-2
  • Molecular Weight : 219.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-cyano-2-fluorobenzenesulfonyl chloride
Inchi Code 1S/C7H3ClFNO2S/c8-13(11,12)7-3-5(4-10)1-2-6(7)9/h1-3H
Inchi Key XLPGJVFDGAPUIH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 4�Light sensitive
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/861347

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P312,P402 + P404,P403 + P233,P410,P411
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-17978_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart